Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981216.2 | XP_008979464.1 | 3 - 4 | 433 | 87 | |
| XM_008981217.2 | XP_008979465.1 | 3 - 4 | 433 | 87 | ||
| XM_008981220.2 | XP_008979468.1 | 3 - 4 | 433 | 87 | ||
| XM_017964596.1 | XP_017820085.1 | 3 - 4 | 433 | 87 | ||
More Information
Set Membership: |
|

ALKAL2
Genomic Map