Assay Details
| Gene Symbol: TIGD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987768.2 | XP_008986016.2 | 1 - 2 | 112 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008987768.2 transcripts.
Set Membership: |
|

TIGD6
Genomic Map