Assay Details
| Gene Symbol: EDNRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003731921.3 | XP_003731969.2 | 5 - 6 | 1185 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDNRB| Gene Symbol: EDNRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003731921.3 | XP_003731969.2 | 5 - 6 | 1185 | 80 | |
Set Membership: |
|