Assay Details
| Gene Symbol: SELENOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974893.1 | XP_017830382.1 | 4 - 5 | 804 | 82 | |
| XM_017974894.1 | XP_017830383.1 | 5 - 6 | 892 | 82 | ||
More Information
Set Membership: |
|

SELENOF
Genomic Map