Assay Details
Gene Symbol: NR3C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257250.2 | NP_001244179.1 | 5 - 6 | 1906 | 66 | |
XM_008986391.1 | XP_008984639.1 | 6 - 7 | 2508 | 66 | ||
XM_008986398.2 | XP_008984646.1 | 6 - 7 | 2113 | 66 | ||
XM_008986403.2 | XP_008984651.1 | 6 - 7 | 2042 | 66 | ||
XM_008986408.2 | XP_008984656.1 | 6 - 7 | 2505 | 66 | ||
XM_008986414.2 | XP_008984662.1 | 6 - 7 | 2073 | 66 | ||
XM_017964545.1 | XP_017820034.1 | 6 - 7 | 2076 | 66 | ||
XM_017964552.1 | XP_017820041.1 | 6 - 7 | 2061 | 66 | ||
XM_017964556.1 | XP_017820045.1 | 6 - 7 | 2484 | 66 | ||
XM_017964560.1 | XP_017820049.1 | 6 - 7 | 2223 | 66 | ||
GenBank mRNA | AY225975.1 | - | 5 - 6 | 1909 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
