Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963821.1 | XP_017819310.1 | 9 - 10 | 1040 | 69 | |
| XM_017963822.1 | XP_017819311.1 | 8 - 9 | 802 | 69 | ||
| XM_017963823.1 | XP_017819312.1 | 8 - 9 | 1037 | 69 | ||
| GenBank mRNA | EF215040.1 | - | 136 | 69 | ||
More Information
Set Membership: |
|

VDAC3
Genomic Map