Assay Details
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257222.1 | NP_001244151.1 | 7 - 8 | 1060 | 69 | |
| XM_008982313.2 | XP_008980561.2 | 7 - 8 | 2925 | 69 | ||
| XM_008982314.2 | XP_008980562.1 | 7 - 8 | 1050 | 69 | ||
| XM_017964597.1 | XP_017820086.1 | 6 - 7 | 3642 | 69 | ||
| XM_017964598.1 | XP_017820087.1 | 7 - 8 | 1072 | 69 | ||
| GenBank mRNA | EF534211.1 | - | 7 - 8 | 1060 | 69 | |
More Information
Set Membership: |
|

CD86
Genomic Map