Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753208.2 | XP_002753254.1 | 2 - 3 | 506 | 78 | |
| XM_009005015.2 | XP_009003263.1 | 2 - 3 | 506 | 78 | ||
More Information
Set Membership: |
|

RPS27L
Genomic Map