Assay Details
| Gene Symbol: LPXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755307.4 | XP_002755353.1 | 5 - 6 | 660 | 65 | |
| XM_009008166.2 | XP_009006414.1 | 4 - 5 | 613 | 65 | ||
More Information
Set Membership: |
|

LPXN
Genomic Map