Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762492.3 | XP_002762538.1 | 7 - 8 | 1040 | 89 | |
| XM_008988649.2 | XP_008986897.1 | 7 - 8 | 1042 | 89 | ||
| XM_008988651.2 | XP_008986899.1 | 6 - 7 | 1561 | 89 | ||
More Information
Set Membership: |
|

SYT5
Genomic Map