Assay Details
| Gene Symbol: USP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986317.1 | XP_008984565.1 | 2202 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP10| Gene Symbol: USP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986317.1 | XP_008984565.1 | 2202 | 93 | ||