Assay Details
| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983306.2 | XP_008981554.1 | 3 - 4 | 3493 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZHX2| Gene Symbol: ZHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983306.2 | XP_008981554.1 | 3 - 4 | 3493 | 91 | |
Set Membership: |
|