Assay Details
| Gene Symbol: TM7SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807364.4 | XP_002807410.4 | 5 - 6 | 755 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM7SF2| Gene Symbol: TM7SF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807364.4 | XP_002807410.4 | 5 - 6 | 755 | 89 | |
Set Membership: |
|