Assay Details
| Gene Symbol: WARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751296.3 | XP_002751342.1 | 4 - 5 | 618 | 61 | |
| XM_002751297.3 | XP_002751343.1 | 4 - 5 | 447 | 61 | ||
| XM_017975058.1 | XP_017830547.1 | 4 - 5 | 614 | 61 | ||
More Information
Set Membership: |
|

WARS2
Genomic Map