Assay Details
| Gene Symbol: ADAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748337.3 | XP_002748383.1 | 2 - 3 | 2444 | 69 | |
| XM_008997118.2 | XP_008995366.1 | 2 - 3 | 2444 | 69 | ||
| XM_008997120.2 | XP_008995368.1 | 2 - 3 | 2444 | 69 | ||
| XM_017972541.1 | XP_017828030.1 | 2 - 3 | 2444 | 69 | ||
| XM_017972542.1 | XP_017828031.1 | 2 - 3 | 2444 | 69 | ||
More Information
Set Membership: |
|

ADAP2
Genomic Map