Assay Details
Gene Symbol: FABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759028.4 | XP_002759074.1 | 2 - 3 | 329 | 65 | |
XM_008982828.2 | XP_008981076.1 | 2 - 3 | 223 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
