Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008987895.2 | XP_008986143.1 | 3 - 4 | 269 | 102 | |
XM_017968131.1 | XP_017823620.1 | 2 - 3 | 202 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
