Assay Details
Gene Symbol: ZNF790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008987895.2 | XP_008986143.1 | 4 - 5 | 365 | 70 | |
XM_017968131.1 | XP_017823620.1 | 3 - 4 | 298 | 70 | ||
XM_017968132.1 | XP_017823621.1 | 4 - 5 | 788 | 70 | ||
XM_017968134.1 | XP_017823623.1 | 3 - 4 | 661 | 70 | ||
XM_017968135.1 | XP_017823624.1 | 2 - 3 | 854 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
