Assay Details
| Gene Symbol: LOC100413008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017972980.1 | XP_017828469.1 | 7 - 8 | 1337 | 99 | |
| XM_017972983.1 | XP_017828472.1 | 7 - 8 | 1337 | 99 | ||
| XM_017972988.1 | XP_017828477.1 | 7 - 8 | 1304 | 99 | ||
| XM_017972989.1 | XP_017828478.1 | 7 - 8 | 1337 | 99 | ||
| XM_017972991.1 | XP_017828480.1 | 7 - 8 | 1337 | 99 | ||
More Information
Set Membership: |
|

LOC100413008
Genomic Map