Assay Details
| Gene Symbol: C10H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754318.3 | XP_002754364.1 | 3 - 4 | 642 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10H14orf180| Gene Symbol: C10H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754318.3 | XP_002754364.1 | 3 - 4 | 642 | 82 | |
Set Membership: |
|