Assay Details
Gene Symbol: ACTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760568.4 | XP_002760614.2 | 1 - 2 | 111 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002760568.4 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
