Assay Details
| Gene Symbol: NPRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758386.4 | XP_002758432.1 | 1 - 2 | 502 | 68 | |
| XM_008981704.2 | XP_008979952.1 | 1 - 2 | 235 | 68 | ||
| XM_017964925.1 | XP_017820414.1 | 1 - 2 | 235 | 68 | ||
| XM_017964926.1 | XP_017820415.1 | 1 - 2 | 507 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017964926.1 transcripts.
Set Membership: |
|

NPRL2
Genomic Map