Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185037.1 | NP_001171966.1 | 3 - 4 | 1272 | 101 | |
| XM_008985811.2 | XP_008984059.1 | 4 - 5 | 1561 | 101 | ||
| XM_008985812.1 | XP_008984060.1 | 4 - 5 | 1776 | 101 | ||
| GenBank mRNA | AJ132343.1 | - | 3 - 4 | 1272 | 101 | |
| EF214844.1 | - | 1 - 2 | 116 | 101 | ||
More Information
Set Membership: |
|

AGT
Genomic Map