Assay Details
| Gene Symbol: HSP90B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752917.3 | XP_002752963.1 | 14 - 15 | 2206 | 86 | |
| GenBank mRNA | EF214869.1 | - | 1 - 2 | 66 | 86 | |
More Information
Set Membership: |
|

HSP90B1
Genomic Map