Assay Details
| Gene Symbol: ERAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997174.2 | XP_008995422.1 | 9 - 10 | 1334 | 86 | |
| GenBank mRNA | EF214932.1 | - | 2 - 3 | 113 | 86 | |
More Information
Set Membership: |
|

ERAL1
Genomic Map