Assay Details
| Gene Symbol: GLRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992548.2 | XP_008990796.1 | 10 | 1768 | 145 | |
| XM_017970179.1 | XP_017825668.1 | 9 | 1564 | 145 | ||
| GenBank mRNA | EF215025.1 | - | 1 | 214 | 145 | |
| EF215318.1 | - | 1 | 208 | 144 | ||
More Information
Set Membership: |
|

GLRB
Genomic Map