Assay Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759986.3 | XP_002760032.1 | 12 - 13 | 1647 | 69 | |
| XM_008984549.1 | XP_008982797.1 | 12 - 13 | 1507 | 69 | ||
| XM_008984550.1 | XP_008982798.1 | 12 - 13 | 1644 | 69 | ||
| XM_008984551.1 | XP_008982799.1 | 12 - 13 | 1644 | 69 | ||
| XM_008984552.1 | XP_008982800.1 | 12 - 13 | 1641 | 69 | ||
| XM_008984553.1 | XP_008982801.1 | 11 - 12 | 1559 | 69 | ||
| XM_008984554.1 | XP_008982802.1 | 10 - 11 | 1226 | 69 | ||
| GenBank mRNA | EF215230.1 | - | 3 - 4 | 195 | 69 | |
More Information
Set Membership: |
|

CLK2
Genomic Map