Assay Details
| Gene Symbol: IL7R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745062.3 | XP_002745108.1 | 2 - 3 | 409 | 64 | |
| GenBank mRNA | EF534213.1 | - | 2 - 3 | 231 | 64 | |
More Information
Set Membership: |
|

IL7R
Genomic Map