Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752907.2 | XP_002752953.1 | 3 - 4 | 382 | 109 | |
| XM_009004489.1 | XP_009002737.1 | 3 - 4 | 382 | 109 | ||
More Information
Set Membership: |
|

WASHC3
Genomic Map