Assay Details
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745573.3 | XP_002745619.1 | 7 - 8 | 1192 | 64 | |
XM_008992942.2 | XP_008991190.1 | 7 - 8 | 745 | 64 | ||
XM_017970406.1 | XP_017825895.1 | 6 - 7 | 1045 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
