Assay Details
| Gene Symbol: PPP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756036.4 | XP_002756082.1 | 7 - 8 | 780 | 88 | |
| XM_009009396.2 | XP_009007644.1 | 6 - 7 | 626 | 88 | ||
| XM_017978844.1 | XP_017834333.1 | 6 - 7 | 678 | 88 | ||
| XM_017978845.1 | XP_017834334.1 | 7 - 8 | 841 | 88 | ||
More Information
Set Membership: |
|

PPP4C
Genomic Map