Assay Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762404.3 | XP_002762450.1 | 3 - 4 | 436 | 70 | |
XM_002762405.3 | XP_002762451.1 | 3 - 4 | 436 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
