Assay Details
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761015.3 | XP_002761061.1 | 2 - 3 | 382 | 67 | |
XM_008985958.2 | XP_008984206.1 | 2 - 3 | 382 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
