Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748008.4 | XP_002748054.1 | 1 | 262 | 62 | |
| XM_008996673.2 | XP_008994921.1 | 1 | 262 | 62 | ||
More Information
Set Membership: |
|

G6PC
Genomic Map