Assay Details
| Gene Symbol: VPS26C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761419.4 | XP_002761465.1 | 1 - 2 | 669 | 100 | |
| XM_003735394.3 | XP_003735442.1 | 1 - 2 | 669 | 100 | ||
More Information
Set Membership: |
|

VPS26C
Genomic Map