Assay Details
| Gene Symbol: SEPT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968530.1 | XP_017824019.1 | 5 - 6 | 899 | 58 | |
| XM_017968531.1 | XP_017824020.1 | 5 - 6 | 899 | 58 | ||
More Information
Set Membership: |
|

SEPT6
Genomic Map