Assay Details
| Gene Symbol: MFHAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979249.2 | XP_008977497.1 | 1 - 2 | 3456 | 66 | |
| XM_008979250.2 | XP_008977498.1 | 1 - 2 | 3456 | 66 | ||
More Information
Set Membership: |
|

MFHAS1
Genomic Map