Assay Details
| Gene Symbol: PTTG1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761489.3 | XP_002761535.1 | 2 - 3 | 373 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTTG1IP| Gene Symbol: PTTG1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761489.3 | XP_002761535.1 | 2 - 3 | 373 | 71 | |
Set Membership: |
|