Assay Details
| Gene Symbol: NKX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753826.4 | XP_002753872.1 | 2 - 3 | 735 | 103 | |
| XM_017977057.1 | XP_017832546.1 | 3 - 4 | 880 | 103 | ||
More Information
Set Membership: |
|

NKX2-1
Genomic Map