Assay Details
| Gene Symbol: SHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968120.1 | XP_017823609.1 | 8 - 9 | 1301 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SHC2| Gene Symbol: SHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968120.1 | XP_017823609.1 | 8 - 9 | 1301 | 66 | |
Set Membership: |
|