Assay Details
| Gene Symbol: LOC100399784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997158.2 | XP_008995406.1 | 14 | 9241 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100399784| Gene Symbol: LOC100399784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997158.2 | XP_008995406.1 | 14 | 9241 | 90 | |
Set Membership: |
|