Assay Details
| Gene Symbol: NECTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982499.2 | XP_008980747.1 | 5 - 6 | 1335 | 92 | |
| XM_008982500.2 | XP_008980748.1 | 5 - 6 | 1335 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NECTIN3| Gene Symbol: NECTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982499.2 | XP_008980747.1 | 5 - 6 | 1335 | 92 | |
| XM_008982500.2 | XP_008980748.1 | 5 - 6 | 1335 | 92 | ||