Assay Details
| Gene Symbol: SERPINB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994061.2 | XP_008992309.2 | 3 - 4 | 493 | 83 | |
| XM_008994062.2 | XP_008992310.1 | 3 - 4 | 1094 | 83 | ||
More Information
Set Membership: |
|

SERPINB9
Genomic Map