Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750064.4 | XP_002750110.1 | 5 - 6 | 1091 | 67 | |
| XM_008999984.2 | XP_008998232.1 | 6 - 7 | 1217 | 67 | ||
| XM_008999985.2 | XP_008998233.1 | 5 - 6 | 1093 | 67 | ||
| XM_008999987.2 | XP_008998235.1 | 5 - 6 | 835 | 67 | ||
| XM_008999988.2 | XP_008998236.1 | 4 - 5 | 609 | 67 | ||
| XM_017974081.1 | XP_017829570.1 | 5 - 6 | 1031 | 67 | ||
More Information
Set Membership: |
|

PTER
Genomic Map