Assay Details
| Gene Symbol: CCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257216.1 | NP_001244145.1 | 1 | 1024 | 62 | |
| GenBank mRNA | AF452614.1 | - | 1 | 1024 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCR5| Gene Symbol: CCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257216.1 | NP_001244145.1 | 1 | 1024 | 62 | |
| GenBank mRNA | AF452614.1 | - | 1 | 1024 | 62 | |
Set Membership: |
|