Assay Details
Gene Symbol: LOC100392681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017973898.1 | XP_017829387.1 | 5 - 6 | 1159 | 70 | |
XM_017973899.1 | XP_017829388.1 | 5 - 6 | 1159 | 70 | ||
XM_017973900.1 | XP_017829389.1 | 4 - 5 | 618 | 70 | ||
XM_017973901.1 | XP_017829390.1 | 5 - 6 | 1159 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
