Assay Details
| Gene Symbol: LOC100392681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017973898.1 | XP_017829387.1 | 7 - 8 | 1383 | 58 | |
| XM_017973899.1 | XP_017829388.1 | 7 - 8 | 1383 | 58 | ||
| XM_017973900.1 | XP_017829389.1 | 6 - 7 | 842 | 58 | ||
More Information
Set Membership: |
|

LOC100392681
Genomic Map