Assay Details
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747787.4 | XP_002747833.1 | 5 - 6 | 1498 | 68 | |
XM_017972182.1 | XP_017827671.1 | 4 - 5 | 1296 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
