Assay Details
| Gene Symbol: ANP32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743118.4 | XP_002743164.1 | 2 - 3 | 639 | 71 | |
| XM_017974963.1 | XP_017830452.1 | 2 - 3 | 639 | 71 | ||
More Information
Set Membership: |
|

ANP32B
Genomic Map