Assay Details
| Gene Symbol: LOC100387916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758966.3 | XP_002759012.1 | 4 - 5 | 1277 | 81 | |
| XM_008982729.2 | XP_008980977.1 | 5 - 6 | 813 | 81 | ||
| XM_008982730.2 | XP_008980978.1 | 4 - 5 | 468 | 81 | ||
| XM_008982735.2 | XP_008980983.1 | 5 - 6 | 813 | 81 | ||
| XM_008982736.2 | XP_008980984.1 | 4 - 5 | 468 | 81 | ||
| XM_017965376.1 | XP_017820865.1 | 5 - 6 | 527 | 81 | ||
| XM_017965377.1 | XP_017820866.1 | 4 - 5 | 456 | 81 | ||
| XM_017965378.1 | XP_017820867.1 | 4 - 5 | 518 | 81 | ||
More Information
Set Membership: |
|

LOC100387916
Genomic Map